Anti-ACTB Mouse Monoclonal Antibody [clone: BA3R]

Supplier: Cell Biolabs
AKR-002
102963-586EA 486.15 USD
102963-586
Anti-ACTB Mouse Monoclonal Antibody [clone: BA3R]
Antibodies
Type: Primary
Antigen: ACTB
Clonality: Monoclonal
Clone: BA3R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Rabbit, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR