Cobalt octoate (6% Co) in mineral spirits

Supplier: Pfaltz & Bauer
Danger

C23740-100G
102963-556EA 164.51 USD
102963-556
Cobalt octoate (6% Co) in mineral spirits
Cobalt octoate
Formula: C₁₆H₃₀CoO₄
Boiling Pt: 179…210 °C
Density: 1.002
Flash Pt: 40 °C
MDL Number: MFCD00072632
CAS Number: 136-52-7
UN: 1268
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR