MELISA-TEK™ Meat Species Kits, Elisa Technologies Inc
Supplier: ELISA TECHNOLOGIES MS
The MELISA-TEK™ Meat Species Kit detects animal species proteins in meat and bone meals and animal feeds.
- Speciation of products containing animal proteins
- Detects only prohibited materials
- Effective with canned and retorted products
The MELISA-TEK™ Meat Species Kit is a method for speciation of products containing animal proteins based on patented monoclonal antibodies sensitive to troponin I, a thermal-stable, muscle-specific antigen
The MELISA-TEK™ Meat Species Kit detects animal species proteins in meat and bone meals and animal feeds. The MELISA Ruminant Kit detects bovine, ovine, and caprine troponin I. The MELISA Pork Kit detects porcine troponin I.
Testing for the presence of the species content in animal feeds is important due to the potential for transmission of TSE. Due to the high processing temperatures used in rendering meat and bone meals, other methods (PCR, conventional species ELISA) are limited in their ability to distinguish species content in animal feed products.
It can determine the species content of thermally processed products including those processed under conditions exceeding 133°C and three bars of pressure. Detects only prohibited materials and not milk or blood and is effective with canned and retorted products.
Ordering information: Microwell ELISA Kits are supplied Ready-To-Use and Refrigerator Stable. Each kit contains the appropriate combination of the following components: 96 sensitized microwells (in a frame with 12 strips of 8 wells each) and the appropriate, calibrated, buffered solutions of conjugated antibodies, controls, diluents, enzyme conjugate, chromogen substrate and buffer, stop solutions, wash concentrates, and instructions for use. Extraction Kit and ELISA kits sold seperately.