Pasteurella Multocida Toxin, List Biological Laboratories
Supplier: List Biological Laboratories, Inc.
156
102946-474EA
683.08
USD
102946-474
Pasteurella Multocida Toxin, List Biological Laboratories
Microbiology Test Systems
Antigen-Antibody Tests
Ag-AK, Seren
Pasteurella multocida toxin is produced by some strains of Pasteurella multocida, a small gram-negative coccobacillus that is pathogenic for many species of animals
Purified Pasteurella multocida toxin has a molecular weight of approximately 146,000 daltons, corresponding to 1,285 amino acids. This toxin can affect the signal transduction pathway of many cells by selective transient activation of a G-protein known as Gqα.