Krazy Glue® Maximum Bond Krazy Glue®
Supplier: Janitorial Supplies
EPIKG48348CO
102945-060EA 4.17 USD
102945-060
Krazy Glue® Maximum Bond Krazy Glue®
Adhesives
Instant-bonding, super-strength glue with no-clog tip.
  • Provides an extra strong, durable, shock-resistant bond
  • Works on ceramic, glass, metal, plastic, wood and rubber
  • Precision tip applicator has an extended, no-clog tip
  • Easy-open, easy to seal hanger cap
  • Larger size and sure-grip panel on the container makes it easier to apply
  • Dries clear
Liquid Super Glue. Clear Color Dried. Clear Color on Application. For adult use.