Anti-CD40 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: HM40-3]

Supplier: Biolegend
102911 102912
102911-BLEA 114.3 USD
102911-BL 102912-BL
Anti-CD40 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: HM40-3]
Antibodies
Alexa Fluor® 647 anti-mouse CD40 [HM40-3]; Isotype: Armenian Hamster IgM; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: HM40-3
Conjugation: Alexa Fluor® 647
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR