tert-Butyl-4-carbamimidoylpiperazine-1-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

115728-1G
102900-220EA 989.23 USD
102900-220
tert-Butyl-4-carbamimidoylpiperazine-1-carboxylate hydrochloride ≥95%
tert-Butyl-4-carbamimidoylpiperazine-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₄O₂
MDL Number: MFCD22421681
CAS Number: 1312935-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR