(3S)-N-(Oxetan-3-yl)piperidin-3-amine ≥95%

Supplier: Matrix Scientific

115950-250MG
102899-978EA 909.23 USD
102899-978
(3S)-N-(Oxetan-3-yl)piperidin-3-amine ≥95%
(3S)-N-(Oxetan-3-yl)piperidin-3-amine

Hazard: Irritant

Formula: C₈H₁₆N₂O
MDL Number: MFCD20233073
CAS Number: 1349699-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR