1,3,5-Tris(Aminomethyl)benzene trihydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Benzene-1,3,5-triyltrimethanamine trihydrochloride

115957-1G 115957-5G 115957-25G
102899-630EA 766.15 USD
102899-630 102899-632 102899-634
1,3,5-Tris(Aminomethyl)benzene trihydrochloride ≥97%
1,3,5-Tris(Aminomethyl)benzene trihydrochloride

Hazard: Irritant

Formula: C₉H₁₈Cl₃N₃
MW: 274.62 g/mol
MDL Number: MFCD22628759
CAS Number: 69146-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR