Methyl-1,4-dihydro-6-methoxy-4-oxo-quinoline-7-carboxylate ≥95%

Supplier: Matrix Scientific

115723-1G
102899-488EA 2581.54 USD
102899-488
Methyl-1,4-dihydro-6-methoxy-4-oxo-quinoline-7-carboxylate ≥95%
Methyl-1,4-dihydro-6-methoxy-4-oxo-quinoline-7-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₄
MDL Number: MFCD18837812
CAS Number: 863786-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR