tert-Butyl-4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate

115321-100MG 115321-250MG 115321-500MG
102899-222EA 1593.85 USD
102899-222 102899-224 102899-226
tert-Butyl-4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₀F₂N₂O₂
MW: 250.29 g/mol
MDL Number: MFCD18909808
CAS Number: 1303973-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR