3-Chloro-N'-cyanobenzene-1-carboximidamide ≥97%

Supplier: Matrix Scientific

115033-500MG 115033-1G
102899-054EA 423.08 USD
102899-054 102899-056
3-Chloro-N'-cyanobenzene-1-carboximidamide ≥97%
3-Chloro-N'-cyanobenzene-1-carboximidamide

Hazard: Irritant

Formula: C₈H₆ClN₃
MDL Number: MFCD24369714
CAS Number: 1432056-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR