4-(Chloromethyl)-2-(methoxymethyl)-1,3-thiazole ≥97%

Supplier: Matrix Scientific

114847-1G
102898-928EA 1104.62 USD
102898-928
4-(Chloromethyl)-2-(methoxymethyl)-1,3-thiazole ≥97%
4-(Chloromethyl)-2-(methoxymethyl)-1,3-thiazole

Hazard: Irritant

Formula: C₆H₈ClNOS
MDL Number: MFCD11178189
CAS Number: 167155-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR