N-(4-Bromobenzyl)oxetan-3-amine ≥95%

Supplier: Matrix Scientific

115849-500MG 115849-1G 115849-5G
102898-480EA 756.92 USD
102898-480 102898-482 102898-484
N-(4-Bromobenzyl)oxetan-3-amine ≥95%
N-(4-Bromobenzyl)oxetan-3-amine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD18849246
CAS Number: 1342610-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR