1,4,5,6-Tetrahydro-4-methyl-3-cyclopentapyrazole methanamine ≥95%

Supplier: Matrix Scientific

115814-250MG 115814-500MG 115814-1G
102898-210EA 1867.69 USD
102898-210 102898-212 102898-214
1,4,5,6-Tetrahydro-4-methyl-3-cyclopentapyrazole methanamine ≥95%
1,4,5,6-Tetrahydro-4-methyl-3-cyclopentapyrazole methanamine

Hazard: Irritant

Formula: C₈H₁₃N₃
MW: 151.21 g/mol
MDL Number: MFCD22566265
CAS Number: 1211596-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR