5-(Boc-amino)thiazole-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-((tert-Butoxycarbonyl)amino)thiazole-2-carboxylic acid

115756-250MG 115756-500MG 115756-1G
102897-408EA 1318.46 USD
102897-408 102897-410 102897-412
5-(Boc-amino)thiazole-2-carboxylic acid ≥97%
5-(Boc-amino)thiazole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₄S
MDL Number: MFCD22415248
CAS Number: 1389264-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR