5-Cbz-1-oxa-5-azaspiro[2.4]heptane

Supplier: Matrix Scientific

113120-1G
102897-330EA 1309.23 USD
102897-330
5-Cbz-1-oxa-5-azaspiro[2.4]heptane
5-Cbz-1-oxa-5-azaspiro[2.4]heptane

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD23105851
CAS Number: 790704-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR