4,7-Dihydro-5H-thieno[2,3-c]pyran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115629-1G
102897-018EA 1787.69 USD
102897-018
4,7-Dihydro-5H-thieno[2,3-c]pyran-3-carboxylic acid ≥95%
4,7-Dihydro-5H-thieno[2,3-c]pyran-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₈O₃S
MDL Number: MFCD17016073
CAS Number: 1169491-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR