tert-Butyl-3,3-difluoro-4-formyl-4-methylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115627-250MG 115627-1G
102897-012EA 2966.15 USD
102897-012 102897-014
tert-Butyl-3,3-difluoro-4-formyl-4-methylpiperidine-1-carboxylate ≥95%
tert-Butyl-3,3-difluoro-4-formyl-4-methylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉F₂NO₃
CAS Number: 1400764-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR