6-Oxa-1-azaspiro[3.3]heptane oxalate ≥95%

Supplier: Matrix Scientific

115592-500MG 115592-1G
102896-942EA 4064.62 USD
102896-942 102896-944
6-Oxa-1-azaspiro[3.3]heptane oxalate ≥95%
6-Oxa-1-azaspiro[3.3]heptane oxalate

Hazard: Irritant

Formula: C₇H₁₁NO₅
MW: 189.17 g/mol
MDL Number: MFCD21364487
CAS Number: 1359655-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR