tert-Butyl-3-(aminomethyl)-4-fluoropiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115459-250MG
115459-500MG
115459-1G
102896-334EA
2746.15
USD
102896-334
102896-336
102896-338
tert-Butyl-3-(aminomethyl)-4-fluoropiperidine-1-carboxylate ≥95%
tert-Butyl-3-(aminomethyl)-4-fluoropiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₁H₂₁FN₂O₂ |
MDL Number:
MFCD18909852 CAS Number: 1303973-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...