4-Methyl-2-thiazolecarbonitrile ≥97%

Supplier: Matrix Scientific

115438-1G 115438-5G
102896-192EA 1264.62 USD
102896-192 102896-194
4-Methyl-2-thiazolecarbonitrile ≥97%
4-Methyl-2-thiazolecarbonitrile

Hazard: Irritant

Formula: C₅H₄N₂S
MW: 124.17 g/mol
MDL Number: MFCD08276856
CAS Number: 100516-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR