(Tributylstannyl)trimethylsilane ≥96%

Supplier: Matrix Scientific

115409-1G 115409-5G 115409-25G
102896-026EA 100 USD
102896-026 102896-028 102896-030
(Tributylstannyl)trimethylsilane ≥96%
(Tributylstannyl)trimethylsilane

Hazard: Irritant

Formula: C₁₅H₃₆SiSn
MDL Number: MFCD00054903
CAS Number: 17955-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR