2-(Acetylamino)-3-(3,4-dimethoxyphenyl)-2-propenoic acid ≥95%

Supplier: Matrix Scientific

115254-500MG 115254-1G
102895-736EA 515.38 USD
102895-736 102895-738
2-(Acetylamino)-3-(3,4-dimethoxyphenyl)-2-propenoic acid ≥95%
2-(Acetylamino)-3-(3,4-dimethoxyphenyl)-2-propenoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₅
MDL Number: MFCD28023548
CAS Number: 75649-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR