2-(1,3-Benzodioxol-5-yl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥97%

Supplier: Matrix Scientific

115045-500MG 115045-1G
102895-274EA 693.85 USD
102895-274 102895-276
2-(1,3-Benzodioxol-5-yl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥97%
2-(1,3-Benzodioxol-5-yl)pyrazolo[1,5-a]pyrazin-4(5H)-one

Hazard: Irritant

Formula: C₁₃H₉N₃O₃
MW: 255.23 g/mol
MDL Number: MFCD17780274
CAS Number: 1255777-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR