Dimethyl-3-cyclopentene-1,1-dicarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Dimethyl cyclopent-3-ene-1,1-dicarboxylate

114720-1G 114720-10G
102894-594EA 253.85 USD
102894-594 102894-596
Dimethyl-3-cyclopentene-1,1-dicarboxylate ≥97%
Dimethyl-3-cyclopentene-1,1-dicarboxylate

Hazard: Irritant

Formula: C₉H₁₂O₄
MW: 184.19 g/mol
MDL Number: MFCD00154460
CAS Number: 84646-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR