5-Fluoro-3-pyridineboronic acid ≥97%

Supplier: Matrix Scientific

114420-1G
102894-000EA 150.77 USD
102894-000
5-Fluoro-3-pyridineboronic acid ≥97%
5-Fluoro-3-pyridineboronic acid

Hazard: Irritant/Refrigerate

Formula: C₅H₅BFNO₂
MDL Number: MFCD07368243
CAS Number: 872041-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR