2-(3-Bromophenoxy)-3-nitropyridine ≥97%

Supplier: Matrix Scientific

114216-1G
102893-580EA 690.77 USD
102893-580
2-(3-Bromophenoxy)-3-nitropyridine ≥97%
2-(3-Bromophenoxy)-3-nitropyridine

Hazard: Irritant

Formula: C₁₁H₇BrN₂O₃
MDL Number: MFCD09948586
CAS Number: 1016802-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR