2-Mercaptopyridine-N-oxide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Mercaptopyridine 1-oxide

114203-1G 114203-10G
102893-550EA 109.23 USD
102893-550 102893-552
2-Mercaptopyridine-N-oxide ≥97%
2-Mercaptopyridine N-oxide

Hazard: Irritant

Formula: C₅H₅NOS
MDL Number: MFCD00006196
CAS Number: 1121-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR