2-(4-Chlorophenyl)-2-morpholinoacetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Chlorophenyl)morpholin-4-ylacetonitrile

114173-1G 114173-10G
102893-500EA 156.92 USD
102893-500 102893-502
2-(4-Chlorophenyl)-2-morpholinoacetonitrile ≥97%
2-(4-Chlorophenyl)-2-morpholinoacetonitrile

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O
MDL Number: MFCD00173106
CAS Number: 33599-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR