Hexafluorocyclotriphosphazene ≥97%
Supplier: Matrix Scientific
113709-1G
102892-652EA
329.23
USD
102892-652
Hexafluorocyclotriphosphazene ≥97%
Hexafluorocyclotriphosphazene
Purity: 97%
CAS: 15599-91-4
MDL Number: MFCD00192400
Molecular Formula: F6N3P3
Molecular Weight: 248,93
Hazard: Irritant
Formula:
F₆N₃P₃ |
MDL Number:
MFCD00192400 CAS Number: 15599-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...