Bis(4-(4-propylcyclohexyl)aniline) sulfuric acid ≥97%

Supplier: Matrix Scientific

115184-500MG 115184-1G
102892-254EA 1040 USD
102892-254 102892-256
Bis(4-(4-propylcyclohexyl)aniline) sulfuric acid ≥97%
Bis(4-(4-propylcyclohexyl)aniline) sulfuric acid

Hazard: Irritant

Formula: C₃₀H₄₈N₂O₄S
MW: 532.79 g/mol
MDL Number: MFCD28023536
CAS Number: 1980039-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR