Bis(4-(4-propylcyclohexyl)aniline) sulfuric acid ≥97%
Supplier: Matrix Scientific
115184-500MG
115184-1G
102892-254EA
1040
USD
102892-254
102892-256
Bis(4-(4-propylcyclohexyl)aniline) sulfuric acid ≥97%
Bis(4-(4-propylcyclohexyl)aniline) sulfuric acid
Hazard: Irritant
Formula:
C₃₀H₄₈N₂O₄S MW: 532.79 g/mol |
MDL Number:
MFCD28023536 CAS Number: 1980039-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...