5-Bromo-2-fluoroisonicotinic acid ≥98%
Supplier: Matrix Scientific
113476-250MG
113476-1G
102892-228EA
300
USD
102892-228
102892-230
5-Bromo-2-fluoroisonicotinic acid ≥98%
5-Bromo-2-fluoroisonicotinic acid
Hazard: Irritant
Formula:
C₆H₃BrFNO₂ MW: 220 g/mol |
MDL Number:
MFCD13185803 CAS Number: 1214377-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...