2-Azido-N-(3-chlorophenyl)acetamide ≥95%

Supplier: Matrix Scientific

115162-500MG 115162-1G
102892-178EA 693.85 USD
102892-178 102892-180
2-Azido-N-(3-chlorophenyl)acetamide ≥95%
2-Azido-N-(3-chlorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇ClN₄O
MW: 210.62 g/mol
MDL Number: MFCD14652257
CAS Number: 215320-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR