1-(3-Tetrahydrofuryl)-1H-pyrazole-4-boronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 1-(Tetrahydrofuran-3-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-(Oxolan-3-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

113284-1G
102891-848EA 1309.23 USD
102891-848
1-(3-Tetrahydrofuryl)-1H-pyrazole-4-boronic acid pinacol ester
1-(3-Tetrahydrofuryl)-1H-pyrazole-4-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₂₁BN₂O₃
MW: 264.13 g/mol
MDL Number: MFCD18383269
CAS Number: 1029715-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR