3-Amino-2-methyl-5-nitropyridine

Supplier: Matrix Scientific

113283-1G
102891-846EA 752.31 USD
102891-846
3-Amino-2-methyl-5-nitropyridine
3-Amino-2-methyl-5-nitropyridine

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD08272071
CAS Number: 51984-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR