cis-4-(Boc-amino)-1-methylcyclohexanol

Supplier: Matrix Scientific

113264-1G
102891-788EA 1740 USD
102891-788
cis-4-(Boc-amino)-1-methylcyclohexanol
cis-4-(Boc-amino)-1-methylcyclohexanol

Hazard: Irritant

Formula: C₁₂H₂₃NO₃
MW: 229.32 g/mol
MDL Number: MFCD21362374
CAS Number: 233764-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR