Linagliptin

Supplier: Matrix Scientific

Synonyms: (R)-8-(3-Aminopiperidin-1-yl)-7-(but-2-yn-1-yl)-3-methyl-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione

113101-1G
102891-484EA 1018.46 USD
102891-484
Linagliptin
Linagliptin

Hazard: Irritant

Formula: C₂₅H₂₈N₈O₂
MDL Number: MFCD14635356
CAS Number: 668270-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR