Anti-CD15 Mouse Monoclonal Antibody (NL557) [clone: MC-480]

Supplier: R&D Systems
NLLC2155R
102887-568EA 808.4 USD
102887-568
Anti-CD15 Mouse Monoclonal Antibody (NL557) [clone: MC-480]
Antibodies
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MC-480
Conjugation: NL557
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR