Anti-CD3D Hamster Monoclonal Antibody [clone: GL-3]

Supplier: R&D Systems
MAB7297
102887-036EA 634.86 USD
102887-036
Anti-CD3D Hamster Monoclonal Antibody [clone: GL-3]
Antibodies
Type: Primary
Antigen: CD3D
Clonality: Monoclonal
Clone: GL-3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG2 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR