Anti-AKR1C1 Mouse Monoclonal Antibody [clone: 85926]

Supplier: R&D Systems
MAB6529
102886-486EA 634.86 USD
102886-486
Anti-AKR1C1 Mouse Monoclonal Antibody [clone: 85926]
Antibodies
Simple Western

Type: Primary
Antigen: AKR1C1
Clonality: Monoclonal
Clone: 85926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR