Anti-NKX3-1 Rat Monoclonal Antibody [clone: 738907]

Supplier: R&D Systems
MAB6475
102886-440EA 620.4 USD
102886-440
Anti-NKX3-1 Rat Monoclonal Antibody [clone: 738907]
Antibodies
Type: Primary
Antigen Symbol: NKX3-1
Antigen Name: NK-3 transcription factor, locus 1 (Drosophila)
Antigen Synonyms: Bax, NKX3.1, NKX3A, Nkx-3.1, bagpipe
Clonality: Monoclonal
Clone: 738907
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR