Anti-S100 A2 Mouse Monoclonal Antibody [clone: 674035]

Supplier: R&D Systems
MAB4870
102885-818EA 620.4 USD
102885-818
Anti-S100 A2 Mouse Monoclonal Antibody [clone: 674035]
Antibodies
Type: Primary
Antigen: S100 A2
Clonality: Monoclonal
Clone: 674035
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR