Anti-IL1R1 Mouse Monoclonal Antibody [clone: 732210]

Supplier: R&D Systems
MAB2691
102885-488EA 634.86 USD
102885-488
Anti-IL1R1 Mouse Monoclonal Antibody [clone: 732210]
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 732210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR