Anti-AFP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 18952]

Supplier: R&D Systems
IC1368P
102884-336EA 562.55 USD
102884-336
Anti-AFP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 18952]
Antibodies
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 18952
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR