Anti-ITGA7 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 334908]

Supplier: R&D Systems
FAB3518N
102882-532EA 634.86 USD
102882-532
Anti-ITGA7 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 334908]
Antibodies
Type: Primary
Antigen: ITGA7
Clonality: Monoclonal
Clone: 334908
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR