Anti-FCRL3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 546828]

Supplier: R&D Systems
FAB3126G
102882-418EA 605.94 USD
102882-418
Anti-FCRL3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 546828]
Antibodies
Type: Primary
Antigen Symbol: FCRL3
Antigen Name: Fc receptor-like 3
Antigen Synonyms: CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Clonality: Monoclonal
Clone: 546828
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR