Anti-TLR2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 383936]

Supplier: R&D Systems
FAB2616N
102882-174EA 591.48 USD
102882-174
Anti-TLR2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 383936]
Antibodies
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: 383936
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR