Anti-TNFSF4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 159403]

Supplier: R&D Systems
FAB10541P
102880-166EA 620.4 USD
102880-166
Anti-TNFSF4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 159403]
Antibodies
Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: 159403
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR